ENA448967762 Enamine

{[(2S,4S)-4-fluoro-1-[(1-methyl-1H-imidazol-2-yl)methyl]pyrrolidin-2-yl]methyl}(methyl)amine

  • MDL number MFCD28505649
  • Cas Number : 1820570-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159452
Availability details for
Quantity
  • Availability
  • Add to Cart