ENA448967772 Enamine

3-(5-amino-2-fluorophenyl)imidazolidine-2,4-dione

  • MDL number MFCD28390265
  • Cas Number : 1803583-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194254
Availability details for
Quantity
  • Availability
  • Add to Cart