ENA448967775 Enamine

3-hydroxypropane-1-sulfonyl fluoride

  • MDL number MFCD28505663
  • Cas Number : 1803600-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195298
Availability details for
Quantity
  • Availability
  • Add to Cart