ENA448967784 Enamine

1-chloro-4-fluoro-2-methoxy-5-methylbenzene

  • MDL number MFCD28505675
  • Cas Number : 211172-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200489
Availability details for
Quantity
  • Availability
  • Add to Cart