ENA448967785 Enamine

3-(3-amino-2-methylphenyl)-1,3-diazinane-2,4-dione hydrochloride

  • MDL number MFCD28505677
  • Cas Number : 1803593-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200694
Availability details for
Quantity
  • Availability
  • Add to Cart