ENA448967787 Enamine

1-(chloromethyl)-2,5-difluoro-4-(trifluoromethyl)benzene

  • MDL number MFCD28505680
  • Cas Number : 1803585-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201467
Availability details for
Quantity
  • Availability
  • Add to Cart