ENA448967790 Enamine

5-fluoro-1-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD28505684
  • Cas Number : 1803589-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202033
Availability details for
Quantity
  • Availability
  • Add to Cart