ENA448967792 Enamine

ethyl N-{[2-(methoxymethyl)phenyl]carbamothioyl}carbamate

  • MDL number MFCD28505686
  • Cas Number : 1803610-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202183
Availability details for
Quantity
  • Availability
  • Add to Cart