ENA448967793 Enamine

3-phenyl-octahydropyrano[4,3-b]pyrrole

  • MDL number MFCD28505688
  • Cas Number : 1803561-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202631
Availability details for
Quantity
  • Availability
  • Add to Cart