ENA448967794 Enamine

1,1,3-trioxo-2,3-dihydro-1lambda6,2-benzothiazole-5-sulfonyl chloride

  • MDL number MFCD28505689
  • Cas Number : 1803601-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203407
Availability details for
Quantity
  • Availability
  • Add to Cart