ENA448967798 Enamine

5-methyl-2,1-benzoxazole

  • MDL number MFCD28505694
  • Cas Number : 566194-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204789
Availability details for
Quantity
  • Availability
  • Add to Cart