ENA448967802 Enamine

methyl 2-cyano-2-cyclobutylacetate

  • MDL number MFCD28505702
  • Cas Number : 1803592-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206061
Availability details for
Quantity
  • Availability
  • Add to Cart