ENA448967810 Enamine

2-fluoro-6-(trifluoromethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD28505710
  • Cas Number : 178049-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206981
Availability details for
Quantity
  • Availability
  • Add to Cart