ENA448967818 Enamine

1-(3,4-dimethoxy-2-methylphenyl)ethan-1-amine

  • MDL number MFCD28505722
  • Cas Number : 1784488-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208213
Availability details for
Quantity
  • Availability
  • Add to Cart