ENA448967820 Enamine

N-(3-chloro-2-hydroxypropyl)pyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD28505725
  • Cas Number : 1803561-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208359
Availability details for
Quantity
  • Availability
  • Add to Cart