ENA448967825 Enamine

2-[2-(cyclopentylamino)acetamido]pent-4-enoic acid hydrochloride

  • MDL number MFCD28505731
  • Cas Number : 1803567-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208619
Availability details for
Quantity
  • Availability
  • Add to Cart