ENA448967829 Enamine

3-(3,5-dimethylphenyl)-3-fluoropyrrolidine hydrochloride

  • MDL number MFCD28505736
  • Cas Number : 1803600-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208995
Availability details for
Quantity
  • Availability
  • Add to Cart