ENA448967832 Enamine

methyl 3-methyl-2-oxo-1,2,3,4-tetrahydroquinoxaline-5-carboxylate

  • MDL number MFCD28505741
  • Cas Number : 1644599-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209454
Availability details for
Quantity
  • Availability
  • Add to Cart