ENA448967834 Enamine

N-[(furan-2-yl)methyl]-3-iodoimidazo[1,2-b]pyridazin-6-amine

  • MDL number MFCD28505743
  • Cas Number : 1803601-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209559
Availability details for
Quantity
  • Availability
  • Add to Cart