ENA448967838 Enamine

2-(1-chloroethyl)-1-phenyl-1H-imidazole hydrochloride

  • MDL number MFCD28505753
  • Cas Number : 1803586-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210301
Availability details for
Quantity
  • Availability
  • Add to Cart