ENA448967844 Enamine

3-chloro-6-(difluoromethoxy)pyridazine

  • MDL number MFCD28502749
  • Cas Number : 1084812-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210843
Availability details for
Quantity
  • Availability
  • Add to Cart