ENA448967846 Enamine

1-[(4-phenylcyclohexyl)sulfanyl]ethan-1-one

  • MDL number MFCD28505762
  • Cas Number : 1803582-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-210921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-210921
Availability details for
Quantity
  • Availability
  • Add to Cart