ENA448967849 Enamine

methyl 4-bromo-2,2-dimethyl-3-oxopentanoate

  • MDL number MFCD28505767
  • Cas Number : 1392512-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211015
Availability details for
Quantity
  • Availability
  • Add to Cart