ENA448967858 Enamine

1-[(3R)-3-aminopiperidin-1-yl]-4,4,4-trifluorobutan-1-one hydrochloride

  • MDL number MFCD28505779
  • Cas Number : 1807938-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211104
Availability details for
Quantity
  • Availability
  • Add to Cart