ENA448967859 Enamine

azetidin-3-yl N-(propan-2-yl)carbamate hydrochloride

  • MDL number MFCD28505781
  • Cas Number : 1803601-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211154
Availability details for
Quantity
  • Availability
  • Add to Cart