ENA448967861 Enamine

2-(1H-pyrrol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD28505784
  • Cas Number : 6662-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211217
Availability details for
Quantity
  • Availability
  • Add to Cart