ENA448967863 Enamine

5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-8-amine hydrochloride

  • MDL number MFCD28505787
  • Cas Number : 1803587-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211278
Availability details for
Quantity
  • Availability
  • Add to Cart