ENA448967864 Enamine

tert-butyl N-(3-{[(3,5-dichlorophenyl)methyl]amino}propyl)carbamate hydrochloride

  • MDL number MFCD28505789
  • Cas Number : 1803604-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211283
Availability details for
Quantity
  • Availability
  • Add to Cart