ENA448967869 Enamine

1-[(2-chloro-5-nitrophenyl)sulfanyl]-N,N-dimethylformamide

  • MDL number MFCD28505803
  • Cas Number : 1803612-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211495
Availability details for
Quantity
  • Availability
  • Add to Cart