ENA448967872 Enamine

5-methanesulfonyl-2-(2-methoxyethoxy)aniline hydrochloride

  • MDL number MFCD28505815
  • Cas Number : 1803612-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211532
Availability details for
Quantity
  • Availability
  • Add to Cart