ENA448967886 Enamine

tert-butyl 8-benzyl-6,6-dimethyl-5-oxa-2,8-diazaspiro[3.5]nonane-2-carboxylate

  • MDL number MFCD28505837
  • Cas Number : 1803602-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211702
Availability details for
Quantity
  • Availability
  • Add to Cart