ENA448967887 Enamine

1-(5,6-dimethylpyridazin-4-yl)methanamine dihydrochloride

  • MDL number MFCD28505838
  • Cas Number : 1803604-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211704
Availability details for
Quantity
  • Availability
  • Add to Cart