ENA448967889 Enamine

tert-butyl 3-[(benzylamino)methyl]-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD28505840
  • Cas Number : 1803589-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211708
Availability details for
Quantity
  • Availability
  • Add to Cart