ENA448967894 Enamine

2-{[(methoxycarbonyl)oxy]methyl}prop-2-enenitrile

  • MDL number MFCD28505848
  • Cas Number : 1803570-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211879
Availability details for
Quantity
  • Availability
  • Add to Cart