ENA448967900 Enamine

2-bromo-1-(2-methoxypyridin-4-yl)ethan-1-one hydrobromide

  • MDL number MFCD28505856
  • Cas Number : 1803589-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211980
Availability details for
Quantity
  • Availability
  • Add to Cart