ENA448967901 Enamine

2-(1-methyl-4-sulfamoyl-1H-pyrrol-2-yl)-2-oxoacetamide

  • MDL number MFCD28505857
  • Cas Number : 1803604-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211989
Availability details for
Quantity
  • Availability
  • Add to Cart