ENA448967912 Enamine

(3R)-1-(thiophene-2-sulfonyl)piperidin-3-amine hydrochloride

  • MDL number MFCD28505870
  • Cas Number : 1807891-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212192
Availability details for
Quantity
  • Availability
  • Add to Cart