ENA448967913 Enamine

tert-butyl N-{3-[(3,4-dimethylcyclohexyl)amino]propyl}carbamate hydrochloride

  • MDL number MFCD28505872
  • Cas Number : 1803606-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212194
Availability details for
Quantity
  • Availability
  • Add to Cart