ENA448967917 Enamine

(1r,4r)-N1,N4-bis(pyridin-2-yl)cyclohexane-1,4-diamine

  • MDL number MFCD28505880
  • Cas Number : 1807941-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212245
Availability details for
Quantity
  • Availability
  • Add to Cart