ENA448967921 Enamine

3-(phenoxymethyl)azetidine hydrochloride

  • MDL number MFCD28505882
  • Cas Number : 1803607-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212289
Availability details for
Quantity
  • Availability
  • Add to Cart