ENA448967925 Enamine

4-{6-[(oxan-4-yl)amino]imidazo[1,2-b]pyridazin-3-yl}benzaldehyde

  • MDL number MFCD26392223
  • Cas Number : 1012345-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212346
Availability details for
Quantity
  • Availability
  • Add to Cart