ENA448967931 Enamine

2-{[2-(pyridin-2-yloxy)ethyl]amino}ethan-1-ol

  • MDL number MFCD28505893
  • Cas Number : 1803602-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212389
Availability details for
Quantity
  • Availability
  • Add to Cart