ENA448967943 Enamine

4-(prop-1-en-1-yl)aniline hydrochloride

  • Cas Number : 1807885-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212941
Availability details for
Quantity
  • Availability
  • Add to Cart