ENA448967946 Enamine

2-(2-{methyl[2-(methylamino)ethyl]amino}ethyl)-2,3-dihydro-1H-isoindole-1,3-dione dihydrochloride

  • MDL number MFCD28505913
  • Cas Number : 1803562-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212966
Availability details for
Quantity
  • Availability
  • Add to Cart