ENA448967949 Enamine

3-(1-chloroethyl)-4-methyl-4H-1,2,4-triazole hydrochloride

  • MDL number MFCD28505916
  • Cas Number : 1803608-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213016
Availability details for
Quantity
  • Availability
  • Add to Cart