ENA448967951 Enamine

2-(aminomethyl)-4,6-dichlorophenol hydrochloride

  • MDL number MFCD28505919
  • Cas Number : 1279870-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213070
Availability details for
Quantity
  • Availability
  • Add to Cart