ENA448967970 Enamine

(4-methylphenyl)methanesulfonamide

  • MDL number MFCD09800673
  • Cas Number : 64732-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200069
Availability details for
Quantity
  • Availability
  • Add to Cart