ENA448967973 Enamine

3-(3-amino-6-fluoro-2-methylphenyl)imidazolidine-2,4-dione

  • MDL number MFCD28505673
  • Cas Number : 1803589-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200136
Availability details for
Quantity
  • Availability
  • Add to Cart