ENA448967981 Enamine

N'-(3-aminophenyl)ethanediamide

  • MDL number MFCD20713267
  • Cas Number : 861333-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200680
Availability details for
Quantity
  • Availability
  • Add to Cart