ENA448967985 Enamine

3-amino-2-(pyridin-3-yl)propan-1-ol

  • MDL number MFCD18802561
  • Cas Number : 62247-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200939
Availability details for
Quantity
  • Availability
  • Add to Cart